accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 683229-61-0

BOOH3CH3CCH3H3CNH3C

CAS RN 683229-61-0

IUPAC Name: 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Synonyms: 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
Molecular Weight (g/mol): 257.14
Molecular Formula: C15H20BNO2
InChi Key: MCCCOWZSGBKRCG-UHFFFAOYSA-N
SMILES: CN1C=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results