accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO(S)NHOSOOO

CAS RN 68399-77-9

IUPAC Name: 4-[(2S)-2-hydroxy-3-sulfonatopropyl]morpholin-4-ium
Synonyms: 4-[(2S)-2-hydroxy-3-sulfonatopropyl]morpholin-4-ium
Molecular Weight (g/mol): 225.26
Molecular Formula: C7H15NO5S
InChi Key: NUFBIAUZAMHTSP-ZETCQYMHSA-N
SMILES: O[C@@H](C[NH+]1CCOCC1)CS([O-])(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)