accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 68401-81-0

IUPAC Name: sodium (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-5-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: ceftizoxime
Molecular Weight (g/mol): 405.38
Molecular Formula: C13H12N5NaO5S2
InChi Key: TVOQBZFDOHZOGA-GIXFXUBFSA-M
SMILES: [Na+].CO\N=C(\C(=O)N[C@H]1[C@H]2SCC=C(N2C1=O)C([O-])=O)C1=CN=C(N)S1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)