accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OO

CAS RN 68480-28-4

IUPAC Name: 3-methylbut-2-en-1-yl formate
Synonyms: 3-methylbut-2-en-1-yl formate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: ZCROFVOAWLRGFY-UHFFFAOYSA-N
SMILES: CC(C)=CCOC=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results