accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH3CNH3CNH3CNFFFSOOOFFFSOOOPd

CAS RN 68569-14-2

IUPAC Name: palladium(2+) tetrakis(acetonitrile) ditrifluoromethanesulfonate
Synonyms: palladium(2+) tetrakis(acetonitrile) ditriflate
Molecular Weight (g/mol): 568.76
Molecular Formula: C10H12F6N4O6PdS2
InChi Key: KXRMHIBEZBOSNG-UHFFFAOYSA-L
SMILES: [Pd++].CC#N.CC#N.CC#N.CC#N.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)