accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BHOOH

CAS RN 68572-87-2

IUPAC Name: (phenanthren-9-yl)boronic acid
Synonyms: phenanthren-9-ylboronic acid
Molecular Weight (g/mol): 222.05
Molecular Formula: C14H11BO2
InChi Key: JCDAUYWOHOLVMH-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results