accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHClCl

CAS RN 68716-47-2

IUPAC Name: (2,4-dichlorophenyl)boronic acid
Synonyms: 2,4-dichlorophenylboronic acid
Molecular Weight (g/mol): 190.81
Molecular Formula: C6H5BCl2O2
InChi Key: QNEGDGPAXKYZHZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results