accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 68758-68-9

H3COOOHOH

CAS RN 68758-68-9

IUPAC Name: ethyl 2-hydroxy-2-(4-hydroxyphenyl)acetate
Synonyms: ethyl 2-hydroxy-2-(4-hydroxyphenyl)acetate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: VLOUFSKXRCPIQR-UHFFFAOYNA-N
SMILES: CCOC(=O)C(O)C1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results