accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SnCH3CH3H

CAS RN 688-73-3

IUPAC Name: tributylstannyl
Synonyms: tributylstannanyl
Molecular Weight (g/mol): 290.06
Molecular Formula: C12H27Sn
InChi Key: PIILXFBHQILWPS-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)CCCC
Boiling Point 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)