accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OOCH3CH2

CAS RN 688-84-6

IUPAC Name: 2-ethylhexyl 2-methylprop-2-enoate
Synonyms: ethylhexyl methacrylate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: WDQMWEYDKDCEHT-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C(C)=C
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)