accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 68800-33-9

CH3OSOOCl

CAS RN 68800-33-9

IUPAC Name: 2-ethoxybenzene-1-sulfonyl chloride
Synonyms: 2-ethoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 220.67
Molecular Formula: C8H9ClO3S
InChi Key: RWURYJNJYODVAL-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results