accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHNHCl

CAS RN 688809-94-1

IUPAC Name: 3-methoxypiperidine hydrochloride
Synonyms: 3-methoxypiperidine hydrochloride
Molecular Weight (g/mol): 151.63
Molecular Formula: C6H14ClNO
InChi Key: ZRDDHBSVIQRILE-UHFFFAOYNA-N
SMILES: Cl.COC1CCCNC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)