accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHOOO

CAS RN 6889-78-7

IUPAC Name: 3-hydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one
Synonyms: 3-hydroxy-2-(4-methoxyphenyl)chromen-4-one
Molecular Weight (g/mol): 268.27
Molecular Formula: C16H12O4
InChi Key: IIBBFGMVMNZMGA-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=C(O)C(=O)C2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results