accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 68915-31-1

IUPAC Name: disodium hydrogen (hydrogen phosphonatooxy)phosphonate
Synonyms: sodium polyphosphate
Molecular Weight (g/mol): 221.94
Molecular Formula: H2Na2O7P2
InChi Key: GYQBBRRVRKFJRG-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results