accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HOOOHO

CAS RN 6893-26-1

IUPAC Name: 2-aminopentanedioic acid
Synonyms: glutaminsaeure
Molecular Weight (g/mol): 147.13
Molecular Formula: C5H9NO4
InChi Key: WHUUTDBJXJRKMK-UHFFFAOYNA-N
SMILES: NC(CCC(O)=O)C(O)=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)