accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

A(a)

CAS RN 68938-01-2

IUPAC Name: 3-(3-bromophenyl)-2-[2-(4-fluorophenyl)acetamido]propanoic acid
Synonyms: 3-(3-bromophenyl)-2-[2-(4-fluorophenyl)acetamido]propanoic acid
Molecular Weight (g/mol): 380.21
Molecular Formula: C17H15BrFNO3
InChi Key: GXZCYECDYOBPGH-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC(Br)=CC=C1)NC(=O)CC1=CC=C(F)C=C1
Physical Form 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)