accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 68971-82-4

H3CH3CCH3OHOHH3CCH3CH3OHH3CH3CCH3OHCH3H3CCH3HOCH3H3CCH3HOCH3CH3CH3OHH3CCH3H3COHCH3CH3H3C

CAS RN 68971-82-4

IUPAC Name: 5,11,17,23,29,35,41,47-octa-tert-butylnonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
Synonyms: 5,11,17,23,29,35,41,47-octa-tert-butylnonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
Molecular Weight (g/mol): 1297.86
Molecular Formula: C88H112O8
InChi Key: OLZFZIXORGGLLS-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(C2)=CC(=C9)C(C)(C)C)=CC(=C8)C(C)(C)C)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)