CAS RN 6899-04-3
IUPAC Name:
2-amino-4-carbamoylbutanoic acid
Synonyms:
glutamin Hgln glutamic acid gamma-amide glutaminsaeure-5-amid
Molecular Weight (g/mol):
146.15
Molecular Formula:
C5H10N2O3
InChi Key:
ZDXPYRJPNDTMRX-UHFFFAOYNA-N
SMILES:
NC(CCC(N)=O)C(O)=O
1
–
2
of
2
results
1
DL-Glutamine, 98%
CAS: 6899-04-3 Molecular Formula: C5H10N2O3 Molecular Weight (g/mol): 146.146 MDL Number: MFCD00065103 InChI Key: ZDXPYRJPNDTMRX-UHFFFAOYSA-N Synonym: dl-glutamine,glutamin,2-amino-4-carbamoylbutanoic acid,dl glutamine,glutamine dl-form,glutamine, dl,l-2-aminoglutaramic acid,2,5-bis azanyl-5-oxidanylidene-pentanoic acid,gamma-glutamine,.gamma.-glutamine PubChem CID: 738 ChEBI: CHEBI:28300 IUPAC Name: 2,5-diamino-5-oxopentanoic acid SMILES: C(CC(=O)N)C(C(=O)O)N
2
DL-glutamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.