accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

A(a)

CAS RN 69011-19-4

IUPAC Name: 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene; trimethylamine
Synonyms: 2-ethylstyrene; divinylbenzene; styrene; trimethylamine
Molecular Weight (g/mol): 425.66
Molecular Formula: C31H39N
InChi Key: HADXLRSCRPYPJJ-UHFFFAOYSA-N
SMILES: CN(C)C.C=CC1=CC=CC=C1.CCC1=CC=CC=C1C=C.C=CC1=CC=CC=C1C=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
Color 
  • (3)
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (9)