accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69011-20-7

IUPAC Name: 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
Synonyms: 2-ethylstyrene; divinylbenzene; styrene
Molecular Weight (g/mol): 366.55
Molecular Formula: C28H30
InChi Key: NWUYHJFMYQTDRP-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1.CCC1=CC=CC=C1C=C.C=CC1=CC=CC=C1C=C
Color 
  • (1)
  • (8)
  • (3)
  • (6)
Quantity 
  • (6)
  • (6)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (1)
  • (1)
  • (8)
  • (3)
  • (6)
  • (6)
  • (6)
  • (6)