accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69014-14-8

H3CNHNSSNNH2H2NNHNN(E/Z)

CAS RN 69014-14-8

IUPAC Name: N''-[4-({[2-(N'-cyano-N''-methylcarbamimidamido)ethyl]sulfanyl}methyl)-1,3-thiazol-2-yl]guanidine
Synonyms: N''-[4-({[2-(N'-cyano-N''-methylcarbamimidamido)ethyl]sulfanyl}methyl)-1,3-thiazol-2-yl]guanidine
Molecular Weight (g/mol): 312.41
Molecular Formula: C10H16N8S2
InChi Key: YDDXVAXDYKBWDX-UHFFFAOYSA-N
SMILES: CN=C(NCCSCC1=CSC(N=C(N)N)=N1)NC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results