accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrOFFFOH

CAS RN 690264-39-2

IUPAC Name: 4-bromo-2-(trifluoromethoxy)phenol
Synonyms: 4-bromo-2-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 257.01
Molecular Formula: C7H4BrF3O2
InChi Key: GYXNTPVMXGBRQW-UHFFFAOYSA-N
SMILES: OC1=C(OC(F)(F)F)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results