accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69047-36-5

IUPAC Name: 1-methyl-1H-indole-7-carbaldehyde
Synonyms: 1-methylindole-7-carbaldehyde
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: JOBIAZYEJOMXEP-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=CC(C=O)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results