accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 690632-04-3

IUPAC Name: 2-(2,3-dihydro-1-benzofuran-5-yl)-4-methyl-1,3-thiazole-5-carboxylic acid
Synonyms: 2-(2,3-dihydro-1-benzofuran-5-yl)-4-methyl-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 261.3
Molecular Formula: C13H11NO3S
InChi Key: DCXMMNGTUXDRND-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC2=C(OCC2)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results