accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 690632-83-8

IUPAC Name: 4-(chloromethyl)-2-(3-chlorophenyl)-1,3-thiazole hydrochloride
Synonyms: 4-(chloromethyl)-2-(3-chlorophenyl)-1,3-thiazole hydrochloride
Molecular Weight (g/mol): 280.59
Molecular Formula: C10H8Cl3NS
InChi Key: CSXLYGPSLQHDEK-UHFFFAOYSA-N
SMILES: Cl.ClCC1=CSC(=N1)C1=CC(Cl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results