CAS RN 69098-15-3
IUPAC Name:
dichloronickel hydrate
Synonyms:
nickel chloride hydrate
Molecular Weight (g/mol):
147.61
Molecular Formula:
Cl2H2NiO
InChi Key:
HGGYAQHDNDUIIQ-UHFFFAOYSA-L
SMILES:
O.Cl[Ni]Cl
1
–
1
of
1
results
1
Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis)
CAS: 69098-15-3 Molecular Formula: Cl2Ni Molecular Weight (g/mol): 129.59 MDL Number: MFCD00149808 InChI Key: QMMRZOWCJAIUJA-UHFFFAOYSA-L Synonym: nickel ii chloride hydrate,nickel chloride hydrate,cl2ni.h2o,nicl2.h2o,nickel 11 chloride hydrate,nickel ii chloride hydrate, puratronic,nickel ii chloride hydrate trace metals basis PubChem CID: 16212333 SMILES: [Cl-].[Cl-].[Ni++]