accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 691-37-2

CH3CH3H2C

CAS RN 691-37-2

IUPAC Name: 4-methylpent-1-ene
Synonyms: 4-methyl-1-pentene polymethylpentene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: WSSSPWUEQFSQQG-UHFFFAOYSA-N
SMILES: CC(C)CC=C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (4)