accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N

CAS RN 6913-92-4

IUPAC Name: 1-benzyl-2,5-dihydro-1H-pyrrole
Synonyms: 1-benzyl-2,5-dihydropyrrole
Molecular Weight (g/mol): 159.23
Molecular Formula: C11H13N
InChi Key: LRFHKHHUKGZIGE-UHFFFAOYSA-N
SMILES: C(N1CC=CC1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results