accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHOO

CAS RN 6914-76-7

IUPAC Name: 1-methylcyclopropane-1-carboxylic acid
Synonyms: 1-methylcyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: DIZKLZKLNKQFGB-UHFFFAOYSA-N
SMILES: CC1(CC1)C(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results