accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

MgCl

CAS RN 6921-34-2

IUPAC Name: magnesium(2+) phenylmethanide chloride
Synonyms: magnesium(2+) phenylmethanide chloride
Molecular Weight (g/mol): 150.89
Molecular Formula: C7H7ClMg
InChi Key: SCEZYJKGDJPHQO-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].[CH2-]C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results