accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6928-85-4

IUPAC Name: 4-methylpiperazin-1-amine
Synonyms: 4-methylpiperazin-1-amine
Molecular Weight (g/mol): 115.18
Molecular Formula: C5H13N3
InChi Key: RJWLLQWLBMJCFD-UHFFFAOYSA-N
SMILES: CN1CCN(N)CC1
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results