accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 693-13-0

IUPAC Name: (propan-2-yl)({[(propan-2-yl)imino]methylidene})amine
Synonyms: diisopropylcarbodiimide N,N-diisopropylcarbodiimide
Molecular Weight (g/mol): 126.2
Molecular Formula: C7H14N2
InChi Key: BDNKZNFMNDZQMI-UHFFFAOYSA-N
SMILES: CC(C)N=C=NC(C)C
Molecular Weight (g/mol) 
  • (7)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (3)