accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHN

CAS RN 693-98-1

IUPAC Name: 2-methyl-1H-imidazole
Synonyms: 2-methylimidazole
Molecular Weight (g/mol): 82.11
Molecular Formula: C4H6N2
InChi Key: LXBGSDVWAMZHDD-UHFFFAOYSA-N
SMILES: CC1=NC=CN1
Molecular Weight (g/mol) 
  • (5)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (6)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)