accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69320-89-4

IUPAC Name: tert-butyl 2-amino-3-methylpentanoate hydrochloride
Synonyms: tert-butyl 2-amino-3-methylpentanoate hydrochloride
Molecular Weight (g/mol): 223.74
Molecular Formula: C10H22ClNO2
InChi Key: IFRYMHOZFAPYPJ-UHFFFAOYNA-N
SMILES: Cl.CCC(C)C(N)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)