accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69365-72-6

NOOONOOONOOOGa

CAS RN 69365-72-6

IUPAC Name: gallium(3+) hydrate trinitrate
Synonyms: gallium nitrate
Molecular Weight (g/mol): 273.75
Molecular Formula: GaH2N3O10
InChi Key: YVFORYDECCQDAW-UHFFFAOYSA-N
SMILES: O.[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (4)
  • (4)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
Grade 
  • (4)
  • (4)
Percent Purity 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (7)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (4)
  • (4)
  • (4)
  • (4)