accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CNCNCNIr

CAS RN 693794-98-8

IUPAC Name: tris(2-(pyridin-2-yl)benzen-1-ide) iridium
Synonyms: tris(2-(pyridin-2-yl)benzen-1-ide) iridium
Molecular Weight (g/mol): 654.8
Molecular Formula: C33H24IrN3
InChi Key: JAZWLEYGTKXZCJ-UHFFFAOYSA-N
SMILES: [Ir].C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results