CAS RN 694-54-2
IUPAC Name:
oxan-2-ol
Synonyms:
oxan-2-ol
Molecular Weight (g/mol):
102.13
Molecular Formula:
C5H10O2
InChi Key:
CELWCAITJAEQNL-UHFFFAOYNA-N
SMILES:
OC1CCCCO1
1
–
2
of
2
results
1
Promotions Available
Tetrahydro-2H-pyran-2-ol, 90%, cyclized form of 5-Hydroxypentanal
Tetrahydro-2H-pyran-2-ol, 90%, cyclized form of 5-Hydroxypentanal
2
Tetrahydro-2H-pyran-2-ol, >90%, TRC
CAS: 694-54-2 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 Synonym: Tetrahydro-2H-pyran-2-ol IUPAC Name: oxan-2-ol SMILES: OC1CCCCO1