accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 694-59-7

IUPAC Name: pyridin-1-ium-1-olate
Synonyms: pyridine N-oxide PY N-oxide
Molecular Weight (g/mol): 95.1
Molecular Formula: C5H5NO
InChi Key: ILVXOBCQQYKLDS-UHFFFAOYSA-N
SMILES: [O-][N+]1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)