accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2FFFF

CAS RN 69411-68-3

IUPAC Name: 3-fluoro-4-(trifluoromethyl)aniline
Synonyms: 3-fluoro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 179.12
Molecular Formula: C7H5F4N
InChi Key: CRRVZRDISHOQQL-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(F)=C1)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results