accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOBrNOO

CAS RN 6942-36-5

IUPAC Name: methyl 2-bromo-5-nitrobenzoate
Synonyms: methyl 2-bromo-5-nitrobenzoate
Molecular Weight (g/mol): 260.04
Molecular Formula: C8H6BrNO4
InChi Key: VSEYYEKRZNRECT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC=C1Br)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results