accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOO

CAS RN 695-95-4

IUPAC Name: methyl 3-oxocyclobutane-1-carboxylate
Synonyms: methyl 3-oxocyclobutane-1-carboxylate
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: IHLHSAIBOSSHQV-UHFFFAOYSA-N
SMILES: COC(=O)C1CC(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results