accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SHNNN

CAS RN 6952-68-7

IUPAC Name: 2H,3H-[1,2,4]triazolo[4,3-a]pyridine-3-thione
Synonyms: 2H-[1,2,4]triazolo[4,3-a]pyridine-3-thione
Molecular Weight (g/mol): 151.19
Molecular Formula: C6H5N3S
InChi Key: ZQMDNIPQCWNIMG-UHFFFAOYSA-N
SMILES: S=C1NN=C2C=CC=CN12
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)