accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNN(E)

CAS RN 696-54-8

IUPAC Name: 4-(nitrosomethylidene)-1,4-dihydropyridine
Synonyms: 4-(nitrosomethylidene)-1H-pyridine
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: XFNAGAQPPRZSFP-UHFFFAOYSA-N
SMILES: O=NC=C1C=CNC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)