accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3O

CAS RN 696-59-3

IUPAC Name: 2,5-dimethoxyoxolane
Synonyms: 2,5-dimethoxyoxolane
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: GFISDBXSWQMOND-UHFFFAOYNA-N
SMILES: COC1CCC(OC)O1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)