accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSH

CAS RN 696-63-9

IUPAC Name: 4-methoxybenzene-1-thiol
Synonyms: 4-methoxythiophenol
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: NIFAOMSJMGEFTQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(S)C=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)