accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHNNOO

CAS RN 6960-45-8

IUPAC Name: 7-nitro-1H-indole-2-carboxylate
Synonyms: 7-nitro-1H-indole-2-carboxylate
Molecular Weight (g/mol): 205.15
Molecular Formula: C9H5N2O4
InChi Key: BIUCOFQROHIAEO-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=CC=CC(=C2N1)[N+]([O-])=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)