accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)NHONOONOO

CAS RN 69632-31-1

IUPAC Name: 3,5-dinitro-N-[(1S)-1-phenylethyl]benzamide
Synonyms: 3,5-dinitro-N-[(1S)-1-phenylethyl]benzamide
Molecular Weight (g/mol): 315.29
Molecular Formula: C15H13N3O5
InChi Key: ABEVDCGKLRIYRW-JTQLQIEISA-N
SMILES: C[C@H](NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results