accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 696602-91-2

BNNHOOHBNNHOOHH2O

CAS RN 696602-91-2

IUPAC Name: bis((pyrimidin-5-yl)boronic acid) hydrate
Synonyms: bis(pyrimidin-5-ylboronic acid) hydrate
Molecular Weight (g/mol): 265.83
Molecular Formula: C8H12B2N4O5
InChi Key: YXMKGFBYXBTOJS-UHFFFAOYSA-N
SMILES: O.OB(O)C1=CN=CN=C1.OB(O)C1=CN=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results