accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69682-13-9

IUPAC Name: 4-tert-butylpiperidine hydrochloride
Synonyms: 4-tert-butylpiperidine hydrochloride
Molecular Weight (g/mol): 177.72
Molecular Formula: C9H20ClN
InChi Key: RWQQDIHTYQYXDX-UHFFFAOYSA-N
SMILES: Cl.CC(C)(C)C1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)