accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NNNBrHN

CAS RN 6974-78-3

IUPAC Name: 8-bromo-7H-purin-6-amine
Synonyms: 8-bromo-7H-purin-6-amine
Molecular Weight (g/mol): 214.03
Molecular Formula: C5H4BrN5
InChi Key: FVXHPCVBOXMRJP-UHFFFAOYSA-N
SMILES: NC1=C2NC(Br)=NC2=NC=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)